5-Methyl-N-carbobenzoxy-L-glutamate ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Z-Glu(OMe)-OHN-Carbobenzoxy-L-glutamic Acid 5-Methyl EsterN-Cbz-L-glutamic Acid 5-Methyl Ester5-Methyl N-Cbz-L-glutamate

0415195GM
100272-650EA 122.92 USD
100272-650
5-Methyl-N-carbobenzoxy-L-glutamate ≥99%, powder
5-Methyl-N-carbobenzoxy-L-glutamate

  • White powder
  • Store at 0-8 °C

Formula: C₁₄H₁₇NO₆
MW: 295.29 g/mol
Melting Pt: 69.6 °C
MDL Number: MFCD00191090
CAS Number: 4652-65-7
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR