D-(+)-Cellobiose ≥98%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Celloseβ-D-Glc-(1→4)-D-Glc4-β-D-glucopyranosyl-D-glueopyranose(2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanalβ-Cellobiose4-O-β-D-Glucopyranosyl-D-glucopyranose

04145925GM
100297-578EA 162.8 USD
100297-578
D-(+)-Cellobiose ≥98%, Powder
D(+)-Cellobiose

  • White powder
  • Store in a cool, dry place

Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Melting Pt: 235…245 °C
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/85/21287485.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR