Ginsenoside Rg1 ≥98% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

020101S5MG
100264-576EA 204 USD
100264-576
Ginsenoside Rg1 ≥98% (by HPLC), powder
Ginsenoside Rg1

  • White powder
  • To be kept in a desiccator

Formula: C₄₂H₇₂O₁₄
MDL Number: MFCD00210293
CAS Number: 22427-39-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/11/21285611.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR