Ginsenoside Rb1 ≥98% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

020105S5MG
100263-372EA 204 USD
100263-372
Ginsenoside Rb1 ≥98% (by HPLC), powder
Ginsenoside Rb1

  • White powder
  • To be kept in a desiccator

Formula: C₅₄H₉₂O₂₃
MDL Number: MFCD00133367
CAS Number: 41753-43-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/51/21285551.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR