D-(+)-Galactosamine hydrochloride ≥98.5%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2R,3R,4R,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

0243421GM
100268-828EA 87.62 USD
100268-828
D-(+)-Galactosamine hydrochloride ≥98.5%, crystals
D(+)-Galactosamine hydrochloride

  • White crystal
  • Store in a cool, dry place

Formula: C₆H₁₄ClNO₅
MW: 215.64 g/mol
Melting Pt: 174…182 °C
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/76/21285876.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR