D-(+)-Fucose ≥98% (by HPLC), crystalline powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
alpha-D-Fucose
0240781GM
100268-770EA
167.64
USD
100268-770
D-(+)-Fucose ≥98% (by HPLC), crystalline powder
D(+)-Fucose
- White crystalline powder
- Store at 0-8 °C
Formula:
C₆H₁₂O₅ Melting Pt: 144…145 °C |
ADR:
, |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...