Fmoc-β-(2-thienyl)-D-alanine ≥98% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (R)-N-Fmoc-2-thienylalanineFmoc-D-Thi-OHFmoc-β-(2-thienyl)-D-Ala-OHFmoc-3-D-Ala(2-thienyl)-OHFmoc-D-2-thienylalanine

0413861GM
100279-150EA 128.15 USD
100279-150
Fmoc-β-(2-thienyl)-D-alanine ≥98% (by HPLC)
Fmoc-β-(2-thienyl)-D-alanine
Formula: C₂₂H₁₉NO₄S
MW: 393.457 g/mol
MDL Number: MFCD00065677
CAS Number: 201532-42-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/65/55/21286555.jpg https://us.vwr.com/stibo/web/std.lang.all/65/54/21286554.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR