Fmoc-3-(1-naphthyl)-D-alanine ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Fmoc-D-1-Nal-OHN-Fmoc-3-(1-naphthyl)-D-alanine

0413321GM
100276-096EA 190.92 USD
100276-096
Fmoc-3-(1-naphthyl)-D-alanine ≥99%, powder
Fmoc-3-(1-naphthyl)-D-alanine

  • White powder
  • Store at 0-8 °C

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
Melting Pt: 182…183 °C
MDL Number: MFCD00191200
CAS Number: 138774-93-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/65/49/21286549.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR