Fmoc-4-methyl-D-phenylalanine ≥98%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
Fmoc-p-Me-D-Phe-OH
, Fmoc-D-Phe(4-Me)-OH
0413551GM
100272-632EA
136
USD
100272-632
Fmoc-4-methyl-D-phenylalanine ≥98%
Fmoc-4-methyl-D-phenylalanine
Formula:
C₂₅H₂₃NO₄ MW: 401.462 g/mol |
MDL Number:
MFCD00270196 CAS Number: 204260-38-8 ADR: , |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...