Fmoc-Lys(Z)-OH ≥98% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Nalpha-Fmoc-Nepsilon-Cbz-L-lysineNalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-carbobenzoxy-L-lysineFmoc-Lys(Z)-OHNalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine

0413135GM
100276-078EA 150.38 USD
100276-078
Fmoc-Lys(Z)-OH ≥98% (by HPLC), powder
Fmoc-Lys(Z)-OH

  • White Powder
  • Store at 0-8 °C

Formula: C₂₉H₃₀N₂O₆
MW: 502.567 g/mol
Melting Pt: 100…130 °C
MDL Number: MFCD00065662
CAS Number: 86060-82-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/71/95/21287195.jpg https://us.vwr.com/stibo/web/std.lang.all/71/94/21287194.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR