Fmoc-3,5-dibromo-D-tyr-OH ≥98% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

0414081GM
100297-566EA 172.62 USD
100297-566
Fmoc-3,5-dibromo-D-tyr-OH ≥98% (by HPLC), powder
Fmoc-3,5-dibromo-D-tyr-OH

  • White powder
  • Store at 0-8 °C

Formula: C₂₄H₁₈NO₅Br₂
Melting Pt: 176…183 °C
MDL Number: MFCD01076139
CAS Number: 204693-22-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/83/21287483.jpg https://us.vwr.com/stibo/web/std.lang.all/74/84/21287484.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR