Fmoc-D-Cha-OH ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Fmoc-β-cyclohexyl-D-Ala-OHN-Fmoc-3-cyclohexyl-D-alanine

0412361GM
100278-920EA 138.62 USD
100278-920
Fmoc-D-Cha-OH ≥99%, crystalline powder
Fmoc-D-Cha-OH

  • White fine crystalline powder
  • Store at 0-8 °C

Formula: C₂₄H₂₇NO₄
MW: 393.483 g/mol
Melting Pt: 131…134 °C
MDL Number: MFCD00151912
CAS Number: 144701-25-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/65/34/21286534.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR