Fmoc-3-(3-benzothienyl)-D-alanine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Fmoc-β-(3-benzothienyl)-D-Ala-OH

0412101GM
100294-530EA 408 USD
100294-530
Fmoc-3-(3-benzothienyl)-D-alanine ≥99%
Fmoc-3-(3-benzothienyl)-D-alanine
Formula: C₂₆H₂₁NO₄S
MW: 443.517 g/mol
MDL Number: MFCD00672563
CAS Number: 177966-61-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/65/30/21286530.jpg https://us.vwr.com/stibo/web/std.lang.all/65/31/21286531.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR