Fmoc-D-Asp-OAll ≥98.5% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Fmoc-D-aspartic acid 1-allyl ester

0412081GM
100275-996EA 235.38 USD
100275-996
Fmoc-D-Asp-OAll ≥98.5% (by HPLC), crystalline powder
Fmoc-D-Asp-OAll

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₂₂H₂₁NO₆
MW: 395.411 g/mol
Melting Pt: 87…124 °C
MDL Number: MFCD01074689
CAS Number: 204246-17-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/62/80/21286280.jpg https://us.vwr.com/stibo/web/std.lang.all/62/79/21286279.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR