Fmoc-ACPC-OH ≥99% (by HPLC), crystalline powder
Supplier: INDOFINE CHEMICAL COMPANY MS
0411801GM
100273-782EA
363.54
USD
100273-782
Fmoc-ACPC-OH ≥99% (by HPLC), crystalline powder
Fmoc-ACPC-OH
- White crystalline powder
- Store at 0-8 °C
Formula:
C₁₉H₁₇NO₄ Melting Pt: 224…226 °C |
MDL Number:
MFCD00190873 CAS Number: 126705-22-5 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...