Fmoc-Dab(Alloc)-OH ≥99% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Fmoc-Dab(Aloc)-OH

0411661GM
100275-970EA 296.85 USD
100275-970
Fmoc-Dab(Alloc)-OH ≥99% (by HPLC), crystalline powder
Fmoc-Dab(Alloc)-OH

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₂₃H₂₄N₂O₆
MW: 424.453 g/mol
Melting Pt: 132…142 °C
MDL Number: MFCD00798634
CAS Number: 204316-32-5
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR