3-Fluoro-4-pyridineboronic acid ≥98%, Powder

Synonyms: (3-Fluoropyridin-4-yl)boronic acid

2638901GM
100287-910EA 394.35 USD
100287-910
3-Fluoro-4-pyridineboronic acid ≥98%, Powder
3-Fluoro-4-pyridineboronic acid

  • White powder
  • Store in a cool, dry place

Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
Melting Pt: 237…239 °C
MDL Number: MFCD03788558
CAS Number: 458532-97-3
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR