4'-(2-Fluorophenyl)propiophenone ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-(2'-Fluorobiphenyl-4-yl)propan-1-one

0935441GM
100271-972EA 109.85 USD
100271-972
4'-(2-Fluorophenyl)propiophenone ≥97%
4'-(2-Fluorophenyl)propiophenone
Formula: C₁₅H₁₃FO
Melting Pt: 76…77 °C
MDL Number: MFCD00236612
CAS Number: 37989-92-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/60/60/21286060.jpg https://us.vwr.com/stibo/web/std.lang.all/60/61/21286061.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR