4-Fluorophenylhydrazinium chloride ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-Fluorophenylhydrazine hydrochloride1-(4-Fluorophenyl)hydrazine hydrochloride(4-Fluorophenyl)hydrazine hydrochloride(4-Fluorphenyl)hydrazin HCl

26102025GM
100272-162EA 104.62 USD
100272-162
4-Fluorophenylhydrazinium chloride ≥97%
4-Fluorophenylhydrazinium chloride
Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
Melting Pt: 250 °C (dec.)
MDL Number: MFCD00012942
CAS Number: 823-85-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/29/21287429.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR