Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-4-Fluoro-α-methylbenzylamine ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: (S)-(-)-1-(4-Fluorophenyl)ethylamine

2638485GM
100287-868EA 324.31 USD InStock
100287-868
(S)-(-)-4-Fluoro-α-methylbenzylamine ≥97%
(S)-(-)-4-Fluoro-α-methylbenzylamine


Formula: C₈H₁₀FN
MW: 139,17 g/mol
Melting Pt: -30 °C
MDL Number: MFCD03092999
CAS Number: 66399-30-2
EINECS: 000-000-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/07/21287007.jpg
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty