4-Fluorobenzylideneacetone ≥97%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
1-(4-Fluorophenyl)but-1-en-3-one
263837500MG
100287-864EA
452.46
USD
100287-864
4-Fluorobenzylideneacetone ≥97%
4-Fluorobenzylideneacetone
Formula:
C₁₀H₉FO |
MDL Number:
MFCD00205144 CAS Number: 1611-38-7 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...