2-Fluoro-6-nitrotoluene, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

2699825GM
100272-486EA 170 USD
100272-486
2-Fluoro-6-nitrotoluene, liquid
2-Fluoro-6-nitrotoluene

  • Clear,yellow liquid
  • Store in a cool, dry place

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 6…8 °C
MDL Number: MFCD00007197
CAS Number: 769-10-8
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/60/75/21286075.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR