3-Fluoro-4-methyl-DL-phenylglycine

Supplier: INDOFINE CHEMICAL COMPANY MS

2637611GM
100298-554EA 319.08 USD
100298-554
3-Fluoro-4-methyl-DL-phenylglycine
3-Fluoro-4-methyl-DL-phenylglycine
Formula: C₉H₁₀FNO₂
MDL Number: MFCD06247587
CAS Number: 261951-76-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/75/25/21287525.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR