8-Fluoro-4-quinolinol

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 8-Fluoroquinolin-4-ol8-Fluoro-4-hydroxyquinoline

262082250MG
100286-416EA 87 USD
100286-416
8-Fluoro-4-quinolinol
8-Fluoro-4-quinolinol
Formula: C₉H₆FNO
MW: 163.15 g/mol
Melting Pt: 278…282 °C
MDL Number: MFCD00272395
CAS Number: 63010-71-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/69/76/21286976.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR