5-Fluorodihydro-6-hydroxy-2,4(1H,3H)pyrimidinedione monohydrate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 5-Fluoro-6-hydroxyhydrouracil monohydrate

269055GM
100278-240EA 224.92 USD
100278-240
5-Fluorodihydro-6-hydroxy-2,4(1H,3H)pyrimidinedione monohydrate ≥98%
5-Fluorodihydro-6-hydroxy-2,4(1H,3H)pyrimidinedione monohydrate
Formula: C₄H₇FN₂O₄
MW: 166.11 g/mol
Melting Pt: >130 °C
MDL Number: MFCD00150905
CAS Number: 37103-91-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/80/21286480.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR