1-(4-Fluorophenyl)-3-(2-hydroxyphenyl)prop-2-en-1-one

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4'-Fluoro-2-hydroxychalcone

2636081GM
100275-438EA 129.46 USD
100275-438
1-(4-Fluorophenyl)-3-(2-hydroxyphenyl)prop-2-en-1-one
1-(4-Fluorophenyl)-3-(2-hydroxyphenyl)prop-2-en-1-one
Formula: C₁₅H₁₁FO₂
MW: 242.25 g/mol
MDL Number: MFCD03788512
CAS Number: 76626-08-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/62/48/21286248.jpg https://us.vwr.com/stibo/web/std.lang.all/62/49/21286249.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR