3-Fluoro-4-hydroxybenzaldehyde ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

213295GM
100285-134EA 119 USD
100285-134
3-Fluoro-4-hydroxybenzaldehyde ≥98%
3-Fluoro-4-hydroxybenzaldehyde
Formula: C₇H₅FO₂
Melting Pt: 121…124 °C
MDL Number: MFCD00016626
CAS Number: 405-05-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/69/43/21286943.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR