4-Fluorobenzoic acid ≥98%

21111100GM
100284-960EA 73.03 USD
100284-960
4-Fluorobenzoic acid ≥98%
4-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Melting Pt: 182…184 °C
MDL Number: MFCD00002530
CAS Number: 456-22-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/69/33/21286933.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR