Ethyl-4-hydroxy-3-methoxycinnamate

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-Hydroxy-3-methoxycinnamic Acid Ethyl EsterEthyl FerulateEthyl 3-(4-hydroxy-3-methoxyphenyl)acrylateFerulic Acid Ethyl Ester

02627510GM
100263-798EA 627.69 USD
100263-798
Ethyl-4-hydroxy-3-methoxycinnamate
Ethyl-4-hydroxy-3-methoxycinnamate
Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
MDL Number: MFCD00009190
CAS Number: 4046-02-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/83/21285583.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR