(-)-Epigallocatechin gallate ≥98% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate(2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoateEGCG

020981S10MG
100262-980EA 117.69 USD
100262-980
(-)-Epigallocatechin gallate ≥98% (by HPLC), powder
(-)-Epigallocatechin gallate

  • Cream Powder
  • Store at 2-8 °C, Away from light

Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
Melting Pt: 220…229 °C
MDL Number: MFCD00075940
CAS Number: 989-51-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/20/21285520.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR