(-)-Epicatechin gallate ≥97.5% (by HPLC), Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (-)-cis-3,3',4',5,7-Pentahydroxyflavane 3-Gallate(2R,3R)-2-(3,4-Dihydroxyphenyl)-5,7-dihydroxychroman-3-yl 3,4,5-trihydroxybenzoate

020978S10MG
100263-470EA 228.85 USD
100263-470
(-)-Epicatechin gallate ≥97.5% (by HPLC), Powder
(-)-Epicatechin gallate

  • Cream Powder
  • Store at 2-8 °C, Away from light

Formula: C₂₂H₁₈O₁₀
MW: 442.38 g/mol
Melting Pt: 257…258 °C
MDL Number: MFCD00075936
CAS Number: 1257-08-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/61/21285561.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR