Emodin ≥99% (by HPLC)

Synonyms: 1,3,8-Trihydroxy-6-methylanthracene-9,10-dione1,3,8-Trihydroxy-6-methylanthraquinone

020453S10MG
100269-706EA 133.75 USD
100269-706
Emodin ≥99% (by HPLC)
Emodin
Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
Melting Pt: 262…266 °C
MDL Number: MFCD00001207
CAS Number: 518-82-1
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR