Ellagic acid ≥97%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2,3,7,8-Tetrahydroxychromeno[5,4,3-cde]chromene-5,10-dione4,4',5,5',6,6'-Hexahydroxydiphenic acid 2,6,2',6'-dilactone

E0015GM
100268-180EA 58.85 USD
100268-180
Ellagic acid ≥97%, powder
Ellagic acid

  • Tan Powder
  • Store in a cool, dry place

Formula: C₁₄H₆O₈
MW: 302.194 g/mol
Melting Pt: >360 °C
MDL Number: MFCD00006914
CAS Number: 476-66-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/56/21285656.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR