11(Z),14(Z),17(Z)-Eicosatrienoic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

102003100MG
100273-370EA 341.31 USD
100273-370
11(Z),14(Z),17(Z)-Eicosatrienoic acid ≥99%
11(Z),14(Z),17(Z)-Eicosatrienoic acid
MDL Number: MFCD00673432
CAS Number: 2091-27-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/33/21286133.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR