n-Dodecyl-β-D-maltoside ≥99%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
Dodecyl-β-D-maltopyranoside
, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
, N-Dodecyl-Beta-Maltoside
, Lauryl-β-D-maltoside
, DDM
0411281GM
100294-488EA
141.23
USD
100294-488
n-Dodecyl-β-D-maltoside ≥99%
n-Dodecyl-β-D-maltoside
Formula:
C₂₄H₄₆O₁₁ MW: 510.6 g/mol |
MDL Number:
MFCD00043012 CAS Number: 69227-93-6 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...