11(Z)-Dodecenoic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

101201100MG
100299-058EA 308.62 USD
100299-058
11(Z)-Dodecenoic acid ≥99%
11(Z)-Dodecenoic acid
MDL Number: MFCD00016758
CAS Number: 65423-25-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/75/33/21287533.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR