Dodecane (mixture of isomers) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

051200100MG
100274-324EA 88 USD
100274-324
Dodecane (mixture of isomers) ≥99%
Dodecane (mixture of isomers)
Formula: C₁₂H₂₆
MW: 170.34 g/mol
MDL Number: MFCD00008969
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/86/21286186.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR