1,3-Dinitro-4-(trifluoromethoxy)benzene ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2,4-Dinitro-1-(trifluoromethoxy)benzene2,4-Dinitro(trifluoromethoxy)benzene

2674825GM
100289-010EA 104.62 USD
100289-010
1,3-Dinitro-4-(trifluoromethoxy)benzene ≥98%
1,3-Dinitro-4-(trifluoromethoxy)benzene
Formula: C₇H₃F₃N₂O₅
MW: 252.11 g/mol
Melting Pt: –21 °C
MDL Number: MFCD00042268
CAS Number: 655-07-2
UN: 2927
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/70/42/21287042.jpg https://us.vwr.com/stibo/web/std.lang.all/70/41/21287041.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR