3-Fluoro-o-xylene ≥98%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
1,2-Dimethyl-3-fluorobenzene
, 1-Fluoro-2,3-dimethylbenzene
0972025GM
100293-532EA
91.15
USD
100293-532
3-Fluoro-o-xylene ≥98%
3-Fluoro-o-xylene
Formula:
C₈H₉F MW: 124.16 g/mol Boiling Pt: 148…152 °C |
MDL Number:
MFCD00000323 CAS Number: 443-82-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...