Flavokawain B ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

02126610MG
100266-102EA 124.23 USD
100266-102
Flavokawain B ≥98%, powder
Flavokawain B

  • Yellow powder
  • Store in a cool, dry place

Formula: C₁₇H₁₆O₄
Melting Pt: 85…87 °C
MDL Number: MFCD00075877
CAS Number: 1775-97-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/02/21285702.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR