3',5'-Dimethoxy-4'-hydroxyacetophenone ≥99%, crystalline powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
1-(3,5-Dimethoxy-4-hydroxyphenyl)ethan-1-one
, 4'-Hydroxy-3',5'-dimethoxyacetophenone
, 1-(4-Hydroxy-3,5-dimethoxyphenyl)ethanone
0260011GM
100266-424EA
26.15
USD
100266-424
3',5'-Dimethoxy-4'-hydroxyacetophenone ≥99%, crystalline powder
3',5'-Dimethoxy-4'-hydroxyacetophenone
- Yellow crystalline powder
- Store in a cool, dry place
Formula:
C₁₀H₁₂O₄ MW: 196.2 g/mol Melting Pt: 125…128 °C |
MDL Number:
MFCD00008748 CAS Number: 2478-38-8 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...