7,8-Dimethoxyflavone ≥98%, needles

Supplier: INDOFINE CHEMICAL COMPANY MS

D024500MG
100265-670EA 210.54 USD
100265-670
7,8-Dimethoxyflavone ≥98%, needles
7,8-Dimethoxyflavone

  • White needles
  • Store in a cool, dry place

Formula: C₁₇H₁₄O₄
Melting Pt: 151…152 °C
MDL Number: MFCD00017463
CAS Number: 65548-54-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/54/21285654.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR