ɑ,ɑ'-Dilaurin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Glycerol ɑ,ɑ'-Dilaurate

32120350MG
100290-264EA 341.31 USD
100290-264
ɑ,ɑ'-Dilaurin ≥99%
alpha,alpha'-Dilaurin
Formula: C₂₇H₅₂O₅
MW: 456.71 g/mol
MDL Number: MFCD00048423
CAS Number: 539-93-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/75/21287075.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR