Gossypetin-3,7,3',4-tetramethylether ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

0212605MG
100266-100EA 1039.62 USD
100266-100
Gossypetin-3,7,3',4-tetramethylether ≥98%
Gossypetin-3,7,3',4-tetramethylether
Formula: C₁₉H₁₈O₈
MDL Number: MFCD00016925
CAS Number: 7380-44-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/01/21285701.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR