4-Methylesculetin ≥98%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 6,7-Dihydroxy-4-methylcoumarin6,7-Dihydroxy-4-methyl-2H-chromen-2-one

D009500MG
100268-090EA 52.31 USD
100268-090
4-Methylesculetin ≥98%, Powder
4-Methylesculetin

  • Cream colored powder
  • Store in a cool, dry place

Formula: C₁₀H₈O₄
MW: 192.17 g/mol
Melting Pt: >280 °C
MDL Number: MFCD00006859
CAS Number: 529-84-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/01/21285801.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR