Sakuranetin ≥99% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

021247S10MG
100266-096EA 274.62 USD
100266-096
Sakuranetin ≥99% (by HPLC), powder
Sakuranetin

  • Cream powder
  • To be kept in a desiccator

Formula: C₁₆H₁₄O₅
Melting Pt: 153…154 °C
MDL Number: MFCD00075651
CAS Number: 2957-21-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/00/21285700.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR