3',4'-Dihydroxyacetophenone ≥90% (by HPLC)

Synonyms: 1-(3,4-Dihydroxyphenyl)ethan-1-oneAcetopyrocatechol

0249561GM
100263-060EA 201.38 USD
100263-060
3',4'-Dihydroxyacetophenone ≥90% (by HPLC)
3',4'-Dihydroxyacetophenone
Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00016439
CAS Number: 1197-09-7
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR