1,3-Difluoropropane ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

2633615GM
100293-988EA 124.23 USD
100293-988
1,3-Difluoropropane ≥97%
1,3-Difluoropropane
Formula: C₃H₆F₂
Boiling Pt: 40…42 °C
MDL Number: MFCD00040969
CAS Number: 462-39-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/72/81/21287281.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR