1-(2,4-Difluorophenyl)-2-thiourea ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2,4-Difluorophenyl)thiourea1-(2,4-Difluorophenyl)thiourea2,4-DifluorophenylthioureaN-(2,4-Difluorophenyl)thiourea

266975GM
100278-208EA 122.92 USD
100278-208
1-(2,4-Difluorophenyl)-2-thiourea ≥97%
1-(2,4-Difluorophenyl)-2-thiourea
Formula: (C₆H₃F₂)NC(S)NH₂
MW: 188.2 g/mol
Melting Pt: 164…167 °C
MDL Number: MFCD00041150
CAS Number: 175277-76-6
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/64/79/21286479.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR