2-Bromo-4'-(difluoromethoxy)acetophenone ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-(Difluoromethoxy)phenacyl bromide

266731GM
100288-952EA 107.23 USD
100288-952
2-Bromo-4'-(difluoromethoxy)acetophenone ≥98%
2-Bromo-4'-(difluoromethoxy)acetophenone
Formula: C₉H₇BrF₂O₂
Melting Pt: 63…68 °C
MDL Number: MFCD00052200
CAS Number: 141134-24-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/38/21287038.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR